Anti-FTH1 Goat Antibody

Supplier: OriGene
ORIGTA305637EA 500 EUR
ORIGTA305637
Anti-FTH1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: FTH1 (ferritin, heavy polypeptide 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR