Anti-STK17B Goat Antibody

Supplier: OriGene
ORIGTA305652EA 502 EUR
ORIGTA305652
Anti-STK17B Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: STK17B (serine/threonine kinase 17b)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR