Imprimer

Anti-USP20 Goat Antibody

Fournisseur: OriGene
ORIGTA305665EA 547 EUR
ORIGTA305665
Anti-USP20 Goat Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 3?g/ml

Type: Primary
Antigen: USP20 (ubiquitin specific peptidase 20)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR