Imprimer

Anti-FAS Goat Antibody

Fournisseur: OriGene
ORIGTA305714EA 547 EUR
ORIGTA305714
Anti-FAS Goat Antibody
Anticorps
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR