Anti-APOL3 Goat antibody

Supplier: OriGene
ORIGTA305719EA 500 EUR
ORIGTA305719
Anti-APOL3 Goat antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: APOL3 (apolipoprotein L, 3)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR