Anti-FHL3 Goat Antibody

Supplier: OriGene
ORIGTA305761EA 502 EUR
ORIGTA305761
Anti-FHL3 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: FHL3 (four and a half LIM domains 3)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR