Imprimer

Anti-FMR1 Goat antibody

Fournisseur: OriGene
ORIGTA305782EA 547 EUR
ORIGTA305782
Anti-FMR1 Goat antibody
Anticorps
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: FMR1 (fragile X mental retardation 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR