Anti-DISP1 Goat Antibody
Supplier: OriGene
ORIGTA305822EA 502 EUR
ORIGTA305822
Anti-DISP1 Goat Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 25?g/ml
Type: Primary
Antigen: DISP1 (dispatched homolog 1 (Drosophila))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat