Anti-APOL6 Goat Antibody

Supplier: OriGene
ORIGTA305871EA 500 EUR
ORIGTA305871
Anti-APOL6 Goat Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: APOL6 (apolipoprotein L, 6)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR