Imprimer

Anti-GALR1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313171EA 564 EUR
ORIGTA313171
Anti-GALR1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: GALR1 (galanin receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR