Anti-TOP2A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313204EA 552 EUR
ORIGTA313204
Anti-TOP2A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: TOP2A (topoisomerase (DNA) II alpha 170kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR