Imprimer

Anti-ZAP70 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313217EA 564 EUR
ORIGTA313217
Anti-ZAP70 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: ZAP70 (zeta-chain (TCR) associated protein kinase 70kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR