Imprimer

Anti-LIMK2 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313235EA 543 EUR
ORIGTA313235
Anti-LIMK2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: LIMK2 (LIM domain kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR