Anti-TFAP2A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313238EA 552 EUR
ORIGTA313238
Anti-TFAP2A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: TFAP2A (transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR