Imprimer

Anti-BCL7A Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313240EA 543 EUR
ORIGTA313240
Anti-BCL7A Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: BCL7A (B-cell CLL/lymphoma 7A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR