Imprimer

Anti-GRK6 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313284EA 543 EUR
ORIGTA313284
Anti-GRK6 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: GRK6 (G protein-coupled receptor kinase 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR