Imprimer

Anti-PTGER3 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313293EA 564 EUR
ORIGTA313293
Anti-PTGER3 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: PTGER3 (prostaglandin E receptor 3 (subtype EP3))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR