Anti-PTGER3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313294EA 552 EUR
ORIGTA313294
Anti-PTGER3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: PTGER3 (prostaglandin E receptor 3 (subtype EP3))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR