Anti-ADRA2A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313316EA 552 EUR
ORIGTA313316
Anti-ADRA2A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:500-1:1000; ELISA: 1:20000

Type: Primary
Antigen: ADRA2A (adrenoceptor alpha 2A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR