Anti-PECAM1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313338EA 552 EUR
ORIGTA313338
Anti-PECAM1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR