Imprimer

Anti-HRH2 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313399EA 543 EUR
ORIGTA313399
Anti-HRH2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: HRH2 (histamine receptor H2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR