Imprimer

Anti-ABCF1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313409EA 564 EUR
ORIGTA313409
Anti-ABCF1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: ABCF1 (ATP-binding cassette, sub-family F (GCN20), member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR