Anti-APBA2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313454EA 552 EUR
ORIGTA313454
Anti-APBA2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: APBA2 (amyloid beta (A4) precursor protein-binding, family A, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR