Anti-ATP5G2 Rabbit Polyclonal Antibody

Supplier: OriGene
TA313489
ORIGTA313489EA 543 EUR
ORIGTA313489
Anti-ATP5G2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: ATP5G2 (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C2 (subunit 9))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR