Imprimer

Anti-CD6 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313575EA 543 EUR
ORIGTA313575
Anti-CD6 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-3000; Immunochemistry: 1:50-100; ELISA: 1:40000

Type: Primary
Antigen: CD6 (CD6 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR