Anti-CDKN2D Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313609EA 552 EUR
ORIGTA313609
Anti-CDKN2D Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: CDKN2D (cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR