Anti-CEBPE Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313611EA 552 EUR
ORIGTA313611
Anti-CEBPE Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CEBPE (CCAAT/enhancer binding protein (C/EBP), epsilon)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR