Anti-COL4A3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313651EA 552 EUR
ORIGTA313651
Anti-COL4A3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: COL4A3 (collagen, type IV, alpha 3 (Goodpasture antigen))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR