Imprimer

Anti-COL4A6 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313655EA 564 EUR
ORIGTA313655
Anti-COL4A6 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: COL4A6 (collagen, type IV, alpha 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR