Imprimer
Anti-AKR1C2 Rabbit Polyclonal Antibody
Fournisseur: OriGene
ORIGTA313748EA 564 EUR
ORIGTA313748
Anti-AKR1C2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000
Type: Primary
Antigen: AKR1C2 (aldo-keto reductase family 1, member C2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human