Anti-ELF5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313809EA 532 EUR
ORIGTA313809
Anti-ELF5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: ELF5 (E74-like factor 5 (ets domain transcription factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR