Anti-EPHB2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313836EA 564 EUR
ORIGTA313836
Anti-EPHB2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: EPHB2 (EPH receptor B2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR