Anti-FANCD2 Rabbit Polyclonal Antibody

Supplier: OriGene
TA313865
ORIGTA313865EA 564 EUR
ORIGTA313865
Anti-FANCD2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: FANCD2 (Fanconi anemia, complementation group D2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR