Imprimer

Anti-FOXD4 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA313874EA 564 EUR
ORIGTA313874
Anti-FOXD4 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:10000

Type: Primary
Antigen: FOXD4 (forkhead box D4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR