Anti-FLT1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313886EA 552 EUR
ORIGTA313886
Anti-FLT1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR