Anti-FSHR Rabbit Polyclonal Antibody

Supplier: OriGene
TA313897
ORIGTA313897EA 528 EUR
ORIGTA313897
Anti-FSHR Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: FSHR (follicle stimulating hormone receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR