Anti-GABBR1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313903EA 552 EUR
ORIGTA313903
Anti-GABBR1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: GABBR1 (gamma-aminobutyric acid (GABA) B receptor, 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR