Anti-GLP1R Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313929EA 564 EUR
ORIGTA313929
Anti-GLP1R Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: GLP1R (glucagon-like peptide 1 receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR