Anti-GNAL Rabbit Polyclonal Antibody

Supplier: OriGene
TA313932
ORIGTA313932EA 543 EUR
ORIGTA313932
Anti-GNAL Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: GNAL (guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR