Anti-GNG5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313935EA 532 EUR
ORIGTA313935
Anti-GNG5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: GNG5 (guanine nucleotide binding protein (G protein), gamma 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR