Anti-GPR31 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313950EA 564 EUR
ORIGTA313950
Anti-GPR31 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: GPR31 (G protein-coupled receptor 31)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR