Anti-MKNK2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313959EA 564 EUR
ORIGTA313959
Anti-MKNK2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: MKNK2 (MAP kinase interacting serine/threonine kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR