Anti-GRIA2 Rabbit Polyclonal Antibody

Supplier: OriGene
TA313965
ORIGTA313965EA 528 EUR
ORIGTA313965
Anti-GRIA2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: GRIA2 (glutamate receptor, ionotropic, AMPA 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR