Anti-GZMH Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA313990EA 564 EUR
ORIGTA313990
Anti-GZMH Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: GZMH (granzyme H (cathepsin G-like 2, protein h-CCPX))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR