Anti-HCFC1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314000EA 552 EUR
ORIGTA314000
Anti-HCFC1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: HCFC1 (host cell factor C1 (VP16-accessory protein))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR