Anti-IL13RA1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314081EA 564 EUR
ORIGTA314081
Anti-IL13RA1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: IL13RA1 (interleukin 13 receptor, alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR