Anti-IRAK1 Rabbit Polyclonal Antibody

Supplier: OriGene
TA314091
ORIGTA314091EA 528 EUR
ORIGTA314091
Anti-IRAK1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: IRAK1 (interleukin-1 receptor-associated kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR