Imprimer

Anti-KCND1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314126EA 543 EUR
ORIGTA314126
Anti-KCND1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: KCND1 (potassium voltage-gated channel, Shal-related subfamily, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR