Anti-KCNS2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314138EA 543 EUR
ORIGTA314138
Anti-KCNS2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: KCNS2 (potassium voltage-gated channel, delayed-rectifier, subfamily S, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR