Anti-LGALS8 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314153EA 552 EUR
ORIGTA314153
Anti-LGALS8 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: LGALS8 (lectin, galactoside-binding, soluble, 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR